Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ethyl hippurate, 98%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Melting Point | 58°C to 60°C |
Quantity | 5g |
Formula Weight | 207.23 |
Percent Purity | 98% |
Chemical Name or Material | Ethyl hippurate |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures