Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl hippurate, 98%

Click to view available options
Quantity:
5g
25g
100g
1499-53-2
C11H13NO3
207.229
MFCD00026890
PTXRQIPIELXJFH-UHFFFAOYSA-N
ethyl hippurate,ethyl n-benzoylglycinate,n-benzoylglycine ethyl ester,ethyl 2-phenylformamido acetate,benzoic amide, n-ethoxycarbonyl methyl,ethylhippurate,ethyl benzoylaminoacetate,acmc-1buir,hippuric acid ethyl ester,maybridge1_006087
226558
ethyl 2-benzamidoacetate
CCOC(=O)CNC(=O)C1=CC=CC=C1
1499-53-2
C11H13NO3
207.229
MFCD00026890
PTXRQIPIELXJFH-UHFFFAOYSA-N
ethyl hippurate,ethyl n-benzoylglycinate,n-benzoylglycine ethyl ester,ethyl 2-phenylformamido acetate,benzoic amide, n-ethoxycarbonyl methyl,ethylhippurate,ethyl benzoylaminoacetate,acmc-1buir,hippuric acid ethyl ester,maybridge1_006087
226558
ethyl 2-benzamidoacetate
CCOC(=O)CNC(=O)C1=CC=CC=C1
Melting Point 58°C to 60°C
Quantity 100g
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material Ethyl hippurate