Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl homovanillate, 97%, Thermo Scientific™

Catalog Number AC189240010
Click to view available options
Quantity:
1 g
60563-13-5
C11H14O4
210.23
MFCD00009180
AWPMWZHWVKXADV-UHFFFAOYSA-N
ethyl homovanillate, ethyl 2-4-hydroxy-3-methoxyphenyl acetate, ethyl 4-hydroxy-3-methoxyphenylacetate, 4-hydroxy-3-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-hydroxy-3-methoxy-, ethyl ester, benzeneacetic acid,4-hydroxy-3-methoxy-, ethyl ester, acmc-20akxa, awpmwzhwvkxadv-uhfffaoysa, ethyl 3-methoxy-4-hydroxy-phenylacetate
108965
ethyl 2-(4-hydroxy-3-methoxyphenyl)acetate
CCOC(=O)CC1=CC=C(O)C(OC)=C1
60563-13-5
C11H14O4
210.23
MFCD00009180
AWPMWZHWVKXADV-UHFFFAOYSA-N
ethyl homovanillate, ethyl 2-4-hydroxy-3-methoxyphenyl acetate, ethyl 4-hydroxy-3-methoxyphenylacetate, 4-hydroxy-3-methoxyphenylacetic acid ethyl ester, benzeneacetic acid, 4-hydroxy-3-methoxy-, ethyl ester, benzeneacetic acid,4-hydroxy-3-methoxy-, ethyl ester, acmc-20akxa, awpmwzhwvkxadv-uhfffaoysa, ethyl 3-methoxy-4-hydroxy-phenylacetate
108965
ethyl 2-(4-hydroxy-3-methoxyphenyl)acetate
CCOC(=O)CC1=CC=C(O)C(OC)=C1
Melting Point 44°C to 47°C
Boiling Point 180°C to 185°C (14.0 mmHg)
Flash Point 113°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula HOC6H3(OCH3)CH2CO2C2H5
Quantity 1 g
Beilstein 10,I,198
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol and acetone
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material Ethyl homovanillate
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 262-298-

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit