Alfa Aesar™ Ethyl hydrogen malonate, 90+%

Manufacturer: Alfa Aesar™ 4469222

Quantity 100g

 View more versions of this product

Catalog No. AA4469222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1071-46-1
    Molecular Formula C5H8O4
    Linear Formula HO2CCH2CO2CH2CH3
    Formula Weight 132.12
    MDL Number MFCD00020490
    Beilstein 1758845
    Chemical Name or Material Ethyl hydrogen malonate

    Specifications

    Formula Weight 132.12
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula HO2CCH2CO2CH2CH3
    Boiling Point 106°C (3mmHg)
    Flash Point >110°C (230°F)
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C5H8O4
    CAS 1071-46-1
    Beilstein 1758845
    Quantity 100g
    Density 1.119g/mL
    Physical Form Liquid
    Refractive Index 1.4295
    MDL Number MFCD00020490
    Health Hazard 1 Warning
    Color Pale yellow
    Chemical Name or Material Ethyl hydrogen malonate
    Assay ≥90%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit