Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl indole-3-carboxylate, 97%

Click to view available options
Quantity:
1g
5g
776-41-0
C11H11NO2
189.214
MFCD00228454
XOUHVMVYFOXTMN-UHFFFAOYSA-N
ethyl indole-3-carboxylate,indole-3-carboxylic acid ethyl ester,1h-indole-3-carboxylic acid ethyl ester,1h-indole-3-carboxylic acid, ethyl ester,indole3carboxylicacidethylester,pubchem7352,lindole-3-ethyl formate,3-ethoxycarbonyl indole,ksc644o3d,3-indolecarboxylicacidethylester
247965
ethyl 1H-indole-3-carboxylate
CCOC(=O)C1=CNC2=CC=CC=C21
776-41-0
C11H11NO2
189.214
MFCD00228454
XOUHVMVYFOXTMN-UHFFFAOYSA-N
ethyl indole-3-carboxylate,indole-3-carboxylic acid ethyl ester,1h-indole-3-carboxylic acid ethyl ester,1h-indole-3-carboxylic acid, ethyl ester,indole3carboxylicacidethylester,pubchem7352,lindole-3-ethyl formate,3-ethoxycarbonyl indole,ksc644o3d,3-indolecarboxylicacidethylester
247965
ethyl 1H-indole-3-carboxylate
CCOC(=O)C1=CNC2=CC=CC=C21
Melting Point 120°C to 124°C
Quantity 5g
Beilstein 147476
Formula Weight 189.21
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Ethyl indole-3-carboxylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning