Alfa Aesar™ Ethyl iodoacetate, 98%

Manufacturer: Alfa Aesar™ B2417722

Quantity 100g

 View more versions of this product

Catalog No. AAB2417722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 623-48-3
    Molecular Formula ICH2CO2CH2CH3
    Formula Weight 214
    MDL Number MFCD00001081
    UN Number UN2927
    Beilstein 741934
    Synonym Iodoacetic acid ethyl ester
    Chemical Name or Material Ethyl iodoacetate

    Specifications

    Formula Weight 214
    TSCA TSCA
    Molecular Formula ICH2CO2CH2CH3
    MDL Number MFCD00001081
    UN Number UN2927
    Beilstein 741934
    Synonym Iodoacetic acid ethyl ester
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H314-H318-H227
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
    RTECS Number AI3575000
    Flash Point 76°C (169°F)
    Boiling Point 179° to 180°C
    Quantity 100g
    CAS 623-48-3
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Ethyl iodoacetate
    Assay 98%

    Description

    Applications
    Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

    Solubility
    Insoluble in water.

    Notes
    Light sensitive. Store in dark. Storage temperature is 2-8°C. Store away from oxidizing agents.

    GHS H Statement
    H300-H314-H318-H227
    Fatal if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit