Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isobutyrylacetate, 95%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7152-15-0
C8H14O3
158.2
MFCD00009198
XCLDSQRVMMXWMS-UHFFFAOYSA-N
ethyl isobutyrylacetate,4-methyl-3-oxo-pentanoic acid ethyl ester,pentanoic acid, 4-methyl-3-oxo-, ethyl ester,ethyl isobutyroylacetate,valeric acid, 4-methyl-3-oxo-, ethyl ester,.gamma.,.gamma.-dimethylacetoacetic acid ethyl ester,beta-keto-gamma-methylvaleric acid ethyl ester,ethyl alpha-isobutyroylacetate,ethylisobutyrylacetate,ethyl iso-butyrylacetate
81583
ethyl 4-methyl-3-oxopentanoate
CCOC(=O)CC(=O)C(C)C
7152-15-0
C8H14O3
158.2
MFCD00009198
XCLDSQRVMMXWMS-UHFFFAOYSA-N
ethyl isobutyrylacetate,4-methyl-3-oxo-pentanoic acid ethyl ester,pentanoic acid, 4-methyl-3-oxo-, ethyl ester,ethyl isobutyroylacetate,valeric acid, 4-methyl-3-oxo-, ethyl ester,.gamma.,.gamma.-dimethylacetoacetic acid ethyl ester,beta-keto-gamma-methylvaleric acid ethyl ester,ethyl alpha-isobutyroylacetate,ethylisobutyrylacetate,ethyl iso-butyrylacetate
81583
ethyl 4-methyl-3-oxopentanoate
CCOC(=O)CC(=O)C(C)C
Infrared Spectrum Authentic
Refractive Index 1.4255 to 1.4275
Quantity 10g
Packaging Glass bottle
Boiling Point 173.0°C
Melting Point -9.0°C
Density 0.9810g/mL
Color Colorless to Yellow
Flash Point 44°C
Specific Gravity 0.981
Assay Percent Range 95%
Linear Formula (CH3)2CHCOCH2CO2C2H5
Formula Weight 158.2
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Ethyl isobutyrylacetate
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning