Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isodehydroacetate, 95%, Acros Organics

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
3385-34-0
C10H12O4
196.2
FBPWNVQUVXSXKS-UHFFFAOYSA-N
ethyl isodehydroacetate,ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate,ethyl isodehydracetate,5-carbethoxy-4,6-dimethyl-2-pyrone,2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester,ethyl 4,6-dimethyl-5-coumalate,2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester,ethylisodehydroacetate,ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate,acmc-20am57
76918
ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
CCOC(=O)C1=C(OC(=O)C=C1C)C
3385-34-0
C10H12O4
196.2
FBPWNVQUVXSXKS-UHFFFAOYSA-N
ethyl isodehydroacetate,ethyl 4,6-dimethyl-2-oxo-2h-pyran-5-carboxylate,ethyl isodehydracetate,5-carbethoxy-4,6-dimethyl-2-pyrone,2h-pyran-5-carboxylic acid, 4,6-dimethyl-2-oxo-, ethyl ester,ethyl 4,6-dimethyl-5-coumalate,2,4-dimethyl-6-oxo-6h-pyran-3-carboxylic acid ethyl ester,ethylisodehydroacetate,ethyl 4,6-dimethyl-2-oxopyran-5-carboxylate,acmc-20am57
76918
ethyl 2,4-dimethyl-6-oxopyran-3-carboxylate
CCOC(=O)C1=C(OC(=O)C=C1C)C
Specific Gravity 1.167
Melting Point 17°C to 18°C
Boiling Point 290°C to 295°C
Flash Point >110°C
Refractive Index 1.5135 to 1.5165
Density 1.1670g/mL
Packaging Glass bottle
Quantity 5g
Assay Percent Range 94% min. (GC)
Beilstein 18,41
Solubility Information Solubility in water: insoluble
Formula Weight 196.2
Percent Purity 95%
Chemical Name or Material Ethyl isodehydroacetate
Show More Show Less

EINECSNumber : 222-195-3