Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Isonipecotate 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1126-09-6
C8H15NO2
157.213
MFCD00006003
RUJPPJYDHHAEEK-UHFFFAOYSA-N
ethyl 4-piperidinecarboxylate,ethyl isonipecotate,piperidine-4-carboxylic acid ethyl ester,ethylpiperidine-4-carboxylate,4-piperidinecarboxylic acid, ethyl ester,isonipecotic acid ethyl ester,4-piperidinecarboxylic acid ethyl ester,4-carbethoxypiperidine,4-carboethoxypiperidine,4-ethoxycarbonyl piperidine
70770
ethyl piperidine-4-carboxylate
CCOC(=O)C1CCNCC1
1126-09-6
C8H15NO2
157.213
MFCD00006003
RUJPPJYDHHAEEK-UHFFFAOYSA-N
ethyl 4-piperidinecarboxylate,ethyl isonipecotate,piperidine-4-carboxylic acid ethyl ester,ethylpiperidine-4-carboxylate,4-piperidinecarboxylic acid, ethyl ester,isonipecotic acid ethyl ester,4-piperidinecarboxylic acid ethyl ester,4-carbethoxypiperidine,4-carboethoxypiperidine,4-ethoxycarbonyl piperidine
70770
ethyl piperidine-4-carboxylate
CCOC(=O)C1CCNCC1
Infrared Spectrum Authentic
Refractive Index 1.4585 to 1.4605
Flash Point 80°C
Specific Gravity 1.02
Packaging Glass bottle
Density 1.0200g/mL
Quantity 100g
Boiling Point 95.0°C to 98.0°C (7.0mmHg)
Color Brown to Colorless
Assay Percent Range 98+%
Beilstein 22, IV, 128
Formula Weight 157.21
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material Ethyl isonipecotate, 98+%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-416-7