Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isovalerate, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
108-64-5
C7H14O2
130.19
MFCD00009203
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl isovalerate,ethyl 3-methylbutyrate,ethylisovalerate,ethyl isopentanoate,isovaleric acid, ethyl ester,butanoic acid, 3-methyl-, ethyl ester,ethyl beta-methylbutyrate,ethyl isovalerianate,3-methylbutyric acid ethyl ester,3-methylbutanoic acid ethyl ester
7945
CHEBI:31571
ethyl 3-methylbutanoate
CCOC(=O)CC(C)C
108-64-5
C7H14O2
130.19
MFCD00009203
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl isovalerate,ethyl 3-methylbutyrate,ethylisovalerate,ethyl isopentanoate,isovaleric acid, ethyl ester,butanoic acid, 3-methyl-, ethyl ester,ethyl beta-methylbutyrate,ethyl isovalerianate,3-methylbutyric acid ethyl ester,3-methylbutanoic acid ethyl ester
7945
CHEBI:31571
ethyl 3-methylbutanoate
CCOC(=O)CC(C)C
Melting Point -99°C
Flash Point 26°C
Boiling Point 131°C to 133°C
Refractive Index 1.3950 to 1.398
Specific Gravity 0.863
Color Colorless to Yellow
Density 0.8630g/mL
Quantity 500mL
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (CH3)2CHCH2CO2C2H5
Beilstein 02, 312
Merck Index 15, 3871
Solubility Information Solubility in water: < 1%. Other solubilities: miscible with alcohol,benzene and ether
Formula Weight 130.19
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl isovalerate, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-602-3

RTECSNumber : NY1504000

TSCA : TSCA