Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl L(-)-lactate, 97%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
687-47-8
C5H10O3
118.13
MFCD00004518
LZCLXQDLBQLTDK-BYPYZUCNSA-N
ethyl l-lactate,ethyl 2s-2-hydroxypropanoate,l-ethyl lactate,l---ethyl lactate,ethyl l--lactate,ethyl s-2-hydroxypropionate,ethyl l---lactate,s-ethyl lactate,propanoic acid, 2-hydroxy-, ethyl ester, 2s
92831
CHEBI:78322
ethyl (2S)-2-hydroxypropanoate
CCOC(=O)C(C)O
687-47-8
C5H10O3
118.13
MFCD00004518
LZCLXQDLBQLTDK-BYPYZUCNSA-N
ethyl l-lactate,ethyl 2s-2-hydroxypropanoate,l-ethyl lactate,l---ethyl lactate,ethyl l--lactate,ethyl s-2-hydroxypropionate,ethyl l---lactate,s-ethyl lactate,propanoic acid, 2-hydroxy-, ethyl ester, 2s
92831
CHEBI:78322
ethyl (2S)-2-hydroxypropanoate
CCOC(=O)C(C)O
Flash Point 46°C
Acidity 0.1% max. (as lactic acid)
Quantity 10L
Packaging Plastic drum
Infrared Spectrum Authentic
Density 1.0340g/mL
Melting Point -26.0°C
Color Undesignated
Specific Rotation Condition − 10.00 (20.00°C neat)
Water 0.3% max. (K.F.)
Specific Gravity 1.034
Refractive Index 1.4100 to 1.4160
Specific Rotation − 10.00
Boiling Point 154.0°C
Assay Percent Range 97%
Linear Formula CH3CH(OH)CO2C2H5
Beilstein 03,264
Fieser 17,135
Merck Index 14,3817
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohols,ketones and esters
Formula Weight 118.13
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl L(-)-lactate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger