Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl m-toluate, 98+%

Click to view available options
Quantity:
25g
100g
120-33-2
C10H12O2
164.204
MFCD00009114
WSJNYOVBJSOQST-UHFFFAOYSA-N
ethyl m-toluate,benzoic acid, 3-methyl-, ethyl ester,ethyl m-methylbenzoate,ethyl-3-methylbenzoate,3-methylbenzoic acid ethyl ester,3-methyl-benzoic acid ethyl ester,m-toluic acid, ethyl ester,ethyl3-methylbenzoate,pubchem15484
67117
ethyl 3-methylbenzoate
CCOC(=O)C1=CC(=CC=C1)C
120-33-2
C10H12O2
164.204
MFCD00009114
WSJNYOVBJSOQST-UHFFFAOYSA-N
ethyl m-toluate,benzoic acid, 3-methyl-, ethyl ester,ethyl m-methylbenzoate,ethyl-3-methylbenzoate,3-methylbenzoic acid ethyl ester,3-methyl-benzoic acid ethyl ester,m-toluic acid, ethyl ester,ethyl3-methylbenzoate,pubchem15484
67117
ethyl 3-methylbenzoate
CCOC(=O)C1=CC(=CC=C1)C
Flash Point 101°C (213°F)
Quantity 100g
Boiling Point 103°C to 105°C (10mmHg)
Beilstein 1863739
Formula Weight 164.21
Percent Purity ≥98%
Chemical Name or Material Ethyl m-toluate