Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl malonyl chloride, 90%, tech., ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
36239-09-5
C5H7ClO3
150.56
KWFADUNOPOSMIJ-UHFFFAOYSA-N
ethyl 3-chloro-3-oxopropionate,ethyl chloroformylacetate,ethyl chloroformyl acetate,propanoic acid, 3-chloro-3-oxo-, ethyl ester,ethyl3-chloro-3-oxopropanoate,ethyl malonylchloride,carbethoxyacetyl chloride,2-ethoxycarbonylacetyl chloride,ethyl 2-chlorocarbonyl acetate,chloroformyl acetic acid ethyl ester
118931
ethyl 3-chloro-3-oxopropanoate
CCOC(=O)CC(=O)Cl
36239-09-5
C5H7ClO3
150.56
KWFADUNOPOSMIJ-UHFFFAOYSA-N
ethyl 3-chloro-3-oxopropionate,ethyl chloroformylacetate,ethyl chloroformyl acetate,propanoic acid, 3-chloro-3-oxo-, ethyl ester,ethyl3-chloro-3-oxopropanoate,ethyl malonylchloride,carbethoxyacetyl chloride,2-ethoxycarbonylacetyl chloride,ethyl 2-chlorocarbonyl acetate,chloroformyl acetic acid ethyl ester
118931
ethyl 3-chloro-3-oxopropanoate
CCOC(=O)CC(=O)Cl
Refractive Index 1.4250 to 1.4310
Specific Gravity 1.17
Boiling Point 79.0°C to 80.0°C (25.0mmHg)
Infrared Spectrum Authentic
Flash Point 67°C
Quantity 100g
Packaging Glass bottle
Density 1.1700g/mL
Color Yellow to Brown
Assay Percent Range 90%
Linear Formula ClCOCH2CO2C2H5
Beilstein 02,582
Solubility Information Solubility in water: reacts
Formula Weight 150.56
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material Ethyl malonyl chloride
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 252-934-5