Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl methacrylate, 99%, stabilized, ACROS Organics™

Click to view available options
Quantity:
25g
500g
1kg
97-63-2
C6H10O2
114.144
MFCD00009161
SUPCQIBBMFXVTL-UHFFFAOYSA-N
ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
7343
ethyl 2-methylprop-2-enoate
CCOC(=O)C(=C)C
97-63-2
C6H10O2
114.144
MFCD00009161
SUPCQIBBMFXVTL-UHFFFAOYSA-N
ethyl methacrylate, ethyl 2-methylacrylate, 2-propenoic acid, 2-methyl-, ethyl ester, methacrylic acid ethyl ester, ethyl 2-methyl-2-propenoate, ethyl 2-methacrylate, rhoplex ac-33, methacrylic acid, ethyl ester, rcra waste number u118, 2-methylacrylic acid, ethyl ester
7343
ethyl 2-methylprop-2-enoate
CCOC(=O)C(=C)C
Stabilizer 10 to 20ppm MEHQ
Color Colorless
Refractive Index 1.4130 to 1.4150 (20°C, 589nm)
Infrared Spectrum Authentic
Quantity 25g
Density 0.9100g/mL
Assay Percent Range 99%
Linear Formula H2C=C(CH3)CO2C2H5
Beilstein 02, 423
Formula Weight 114.14
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl methacrylate, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 202-597-5

RTECSNumber : OZ4550000

TSCA : TSCA