Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl methoxyacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
50g
Packaging:
Glass bottle
3938-96-3
C5H10O3
118.132
MFCD00009176
JLEKJZUYWFJPMB-UHFFFAOYSA-N
ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
77544
ethyl 2-methoxyacetate
CCOC(=O)COC
3938-96-3
C5H10O3
118.132
MFCD00009176
JLEKJZUYWFJPMB-UHFFFAOYSA-N
ethyl methoxyacetate, acetic acid, methoxy-, ethyl ester, 2-methoxy ethyl acetate, ethyl-3-oxabutanoate, methoxyacetic acid ethyl ester, methoxy acetic acid ethyl ester, ethyl-metoxyacetate, methylglycol acetate, ethyl methyloxy acetate, acmc-1cofg
77544
ethyl 2-methoxyacetate
CCOC(=O)COC
Quantity 50g
Packaging Glass bottle
Density 1.0000g/mL
Color Colorless
Infrared Spectrum Authentic
Refractive Index 1.3985 to 1.4015 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula CH3OCH2CO2C2H5
Beilstein 03, 236
Formula Weight 118.13
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Ethyl methoxyacetate, 98%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 223-521-7