Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (methylthio)acetate, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
4455-13-4
C5H10O2S
134.193
MFCD00009182
MDIAKIHKBBNYHF-UHFFFAOYSA-N
ethyl methylthio acetate,ethyl 2-methylthio acetate,acetic acid, methylthio-, ethyl ester,ethyl 2-methylthio-acetate,ethyl 2-methylsulfanyl acetate,unii-6148qqn25t,ethyl alpha-methylthio acetate,methylthio acetic acid ethyl ester,ethyl .alpha. methylthio acetate,methylsulfanyl-acetic acid ethyl ester
78199
ethyl 2-methylsulfanylacetate
CCOC(=O)CSC
4455-13-4
C5H10O2S
134.193
MFCD00009182
MDIAKIHKBBNYHF-UHFFFAOYSA-N
ethyl methylthio acetate,ethyl 2-methylthio acetate,acetic acid, methylthio-, ethyl ester,ethyl 2-methylthio-acetate,ethyl 2-methylsulfanyl acetate,unii-6148qqn25t,ethyl alpha-methylthio acetate,methylthio acetic acid ethyl ester,ethyl .alpha. methylthio acetate,methylsulfanyl-acetic acid ethyl ester
78199
ethyl 2-methylsulfanylacetate
CCOC(=O)CSC
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4577 to 1.4597 (20°C, 589nm)
Quantity 100g
Color Colorless to Yellow
Density 1.04g/mL
Assay Percent Range 95%
Linear Formula CH3SCH2CO2C2H5
Formula Weight 134.19
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Ethyl (methylthio)acetate, 95%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-700-2