Alfa Aesar™ Ethyl (methylthio)acetate, 98%

Manufacturer: Alfa Aesar™ B2428506

Quantity 5g

 View more versions of this product

Catalog No. AAB2428506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4455-13-4
    Molecular Formula CH3SCH2CO2CH2CH3
    Formula Weight 134.2
    MDL Number MFCD00009182
    UN Number UN3272
    Beilstein 1744999
    Synonym (Methylthio)acetic acid ethyl ester
    Chemical Name or Material Ethyl (methylthio)acetate

    Specifications

    Formula Weight 134.2
    Molecular Formula CH3SCH2CO2CH2CH3
    MDL Number MFCD00009182
    CAS 4455-13-4
    UN Number UN3272
    Beilstein 1744999
    Synonym (Methylthio)acetic acid ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H319-H335
    Flammable liquid and vapour.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 59°C (138°F)
    Odor Characteristic
    Boiling Point 176°C
    Quantity 5g
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    Chemical Name or Material Ethyl (methylthio)acetate
    Assay 98%

    Description

    Applications
    Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.

    Solubility
    Miscible with alcohol. Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents and strong alkalis.

    GHS H Statement
    H226-H319-H335
    Flammable liquid and vapour.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit