Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl (methylthio)acetate, 98%

Click to view available options
Quantity:
5g
25g
100g
4455-13-4
C5H10O2S
134.193
MFCD00009182
MDIAKIHKBBNYHF-UHFFFAOYSA-N
ethyl methylthio acetate,ethyl 2-methylthio acetate,acetic acid, methylthio-, ethyl ester,ethyl 2-methylthio-acetate,ethyl 2-methylsulfanyl acetate,unii-6148qqn25t,ethyl alpha-methylthio acetate,methylthio acetic acid ethyl ester,ethyl .alpha. methylthio acetate,methylsulfanyl-acetic acid ethyl ester
78199
ethyl 2-methylsulfanylacetate
CCOC(=O)CSC
4455-13-4
C5H10O2S
134.193
MFCD00009182
MDIAKIHKBBNYHF-UHFFFAOYSA-N
ethyl methylthio acetate,ethyl 2-methylthio acetate,acetic acid, methylthio-, ethyl ester,ethyl 2-methylthio-acetate,ethyl 2-methylsulfanyl acetate,unii-6148qqn25t,ethyl alpha-methylthio acetate,methylthio acetic acid ethyl ester,ethyl .alpha. methylthio acetate,methylsulfanyl-acetic acid ethyl ester
78199
ethyl 2-methylsulfanylacetate
CCOC(=O)CSC
Applications
Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong alkalis.
Odor Characteristic
Boiling Point 176°C
Flash Point 59°C (138°F)
Quantity 100g
UN Number UN3272
Beilstein 1744999
Formula Weight 134.2
Percent Purity 98%
Chemical Name or Material Ethyl (methylthio)acetate

GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.