Alfa Aesar™ Ethyl N-(4-bromophenyl)oxamate

Manufacturer: Alfa Aesar™ H5059803

Quantity 1g

 View more versions of this product

Catalog No. AAH5059803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 24451-15-8
    Molecular Formula C10H10BrNO3
    Formula Weight 272.1
    MDL Number MFCD00728542
    Synonym N-(4-Bromophenyl)oxamic acid ethyl ester; Ethyl (4-bromoanilino)oxoacetate
    Chemical Name or Material Ethyl N-(4-bromophenyl)oxamate

    Specifications

    Formula Weight 272.1
    Molecular Formula C10H10BrNO3
    MDL Number MFCD00728542
    CAS 24451-15-8
    Synonym N-(4-Bromophenyl)oxamic acid ethyl ester; Ethyl (4-bromoanilino)oxoacetate
    Quantity 1g
    Chemical Name or Material Ethyl N-(4-bromophenyl)oxamate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit