Alfa Aesar™ Ethyl N-hydroxyacetimidate, 97%

Manufacturer: Alfa Aesar™ A1100622

Quantity 100g

 View more versions of this product

Catalog No. AAA1100622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10576-12-2
    Molecular Formula C4H9NO2
    Formula Weight 103.12
    MDL Number MFCD00002114
    Beilstein 1700260
    Synonym Ethyl acetohydroximate
    Chemical Name or Material Ethyl N-hydroxyacetimidate

    Specifications

    Formula Weight 103.12
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Molecular Formula C4H9NO2
    MDL Number MFCD00002114
    CAS 10576-12-2
    Beilstein 1700260
    Melting Point 23° to 25°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Boiling Point 55° to 58°C (6mmHg)
    Recommended Storage 2°C to 8°C
    Synonym Ethyl acetohydroximate
    Flash Point 77°C (171°F)
    Quantity 100g
    Chemical Name or Material Ethyl N-hydroxyacetimidate
    Assay 97%

    Description

    Applications
    Ethyl N-hydroxyacetimidate was used in the synthesis of O-acyl-and O-nitrophenylhydroxylamines, hydroxamic acid ethoxycarbonylhydrazides and ethyl O-(2,4-dinitrophenyl)acetohydroxamate.

    Solubility
    Decompose in water.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    2°C to 8°C

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit