Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl N-hydroxyacetimidate, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
10576-12-2
C4H9NO2
103.12
MFCD00002114
QWKAVVNRCKPKNM-PLNGDYQASA-N
ethyl n-hydroxyacetimidate,n-hydroxyethanimidic acid, ethyl ester,ethyl acetohydroximate,ethyl n-hydroxyethanimidate,e-ethyl n-hydroxyethanimidate,e-ethyl n-hydroxyacetimidate,ethyl 1e-n-hydroxyethanimidate,snx`lfelmmpqh,ethyl acethydroxamate,1-ethoxyethanone oxime
6386647
ethyl (1Z)-N-hydroxyethanimidate
CCOC(=NO)C
10576-12-2
C4H9NO2
103.12
MFCD00002114
QWKAVVNRCKPKNM-PLNGDYQASA-N
ethyl n-hydroxyacetimidate,n-hydroxyethanimidic acid, ethyl ester,ethyl acetohydroximate,ethyl n-hydroxyethanimidate,e-ethyl n-hydroxyethanimidate,e-ethyl n-hydroxyacetimidate,ethyl 1e-n-hydroxyethanimidate,snx`lfelmmpqh,ethyl acethydroxamate,1-ethoxyethanone oxime
6386647
ethyl (1Z)-N-hydroxyethanimidate
CCOC(=NO)C
Melting Point 23.0°C to 25.0°C
Boiling Point 55.0°C to 58.0°C (6.0mmHg)
Infrared Spectrum Authentic
Refractive Index 1.4325 to 1.4345
Flash Point 77°C
Color Colorless
Packaging Glass bottle
Quantity 10g
Assay Percent Range 96% min. (GC)
Linear Formula CH3C(=NOH)OC2H5
Solubility Information Solubility in water: may decompose.
Formula Weight 103.12
Physical Form Liquid After Melting
Percent Purity 97%
Chemical Name or Material Ethyl N-hydroxyacetimidate, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 234-165-7

TSCA : TSCA