Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Nicotinate, Stabilized 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
614-18-6
C8H9NO2
151.165
MFCD00006389
XBLVHTDFJBKJLG-UHFFFAOYSA-N
ethyl nicotinate,ethyl nicotinoate,nicotinic acid ethyl ester,ethyl 3-pyridinecarboxylate,3-pyridinecarboxylic acid, ethyl ester,ethylnicotinate,nicotinic acid, ethyl ester,mucotherm,nicaethan,nikethan
69188
ethyl pyridine-3-carboxylate
CCOC(=O)C1=CN=CC=C1
614-18-6
C8H9NO2
151.165
MFCD00006389
XBLVHTDFJBKJLG-UHFFFAOYSA-N
ethyl nicotinate,ethyl nicotinoate,nicotinic acid ethyl ester,ethyl 3-pyridinecarboxylate,3-pyridinecarboxylic acid, ethyl ester,ethylnicotinate,nicotinic acid, ethyl ester,mucotherm,nicaethan,nikethan
69188
ethyl pyridine-3-carboxylate
CCOC(=O)C1=CN=CC=C1
Quantity 100g
Color Colorless to Yellow
Water 0.5% max. (K.F.)
Refractive Index 1.5019 to 1.5040 (20°C, 589nm)
Density 1.1000g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Beilstein 22, 39
Formula Weight 151.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl nicotinate, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 210-370-7

TSCA : TSCA

Recommended Storage : This product becomes brown during storage