Alfa Aesar™ Ethyl nipecotate, 97%

Manufacturer: Alfa Aesar™ B2415514

Quantity 25g

 View more versions of this product

Catalog No. AAB2415514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5006-62-2
    Molecular Formula C8H15NO2
    Formula Weight 157.21
    MDL Number MFCD00005991
    Synonym Ethyl piperidine-3-carboxylate; Nipecotic acid ethyl ester
    Chemical Name or Material Ethyl nipecotate

    Specifications

    Formula Weight 157.21
    Flash Point 90°C (195°F)
    Molecular Formula C8H15NO2
    MDL Number MFCD00005991
    CAS 5006-62-2
    Synonym Ethyl piperidine-3-carboxylate; Nipecotic acid ethyl ester
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Boiling Point 102° to 104°C (7mmHg)
    Quantity 25g
    Chemical Name or Material Ethyl nipecotate
    Assay 97%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    2° to 8°C

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit