Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl nipecotate, 97%

Click to view available options
Quantity:
25g
100g
5006-62-2
C8H15NO2
157.213
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl nipecotate,ethyl 3-piperidinecarboxylate,nipecotic acid ethyl ester,3-piperidinecarboxylic acid ethyl ester,ethylpiperidine-3-carboxylate,3-ethoxycarbonyl piperidine,3-piperidinecarboxylic acid, ethyl ester,piperidine-3-carboxylic acid ethyl ester,ethylnipecotate,a-ethyl nipecotate
98969
ethyl piperidine-3-carboxylate
CCOC(=O)C1CCCNC1
5006-62-2
C8H15NO2
157.213
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl nipecotate,ethyl 3-piperidinecarboxylate,nipecotic acid ethyl ester,3-piperidinecarboxylic acid ethyl ester,ethylpiperidine-3-carboxylate,3-ethoxycarbonyl piperidine,3-piperidinecarboxylic acid, ethyl ester,piperidine-3-carboxylic acid ethyl ester,ethylnipecotate,a-ethyl nipecotate
98969
ethyl piperidine-3-carboxylate
CCOC(=O)C1CCCNC1
Flash Point 90°C (195°F)
Boiling Point 102°C to 104°C (7mmHg)
Quantity 100g
Formula Weight 157.21
Percent Purity 97%
Chemical Name or Material Ethyl nipecotate

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C