Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl nonanoate, 97%

Click to view available options
Quantity:
100mL
500mL
123-29-5
C11H22O2
186.295
MFCD00009570
BYEVBITUADOIGY-UHFFFAOYSA-N
ethyl pelargonate,nonanoic acid, ethyl ester,ethyl nonylate,wine ether,nonanoic acid ethyl ester,natural,ethyl n-nonanoate,unii-ksh683s98j,fema no. 2447,pelargonic acid ethyl ester
31251
CHEBI:87501
ethyl nonanoate
CCCCCCCCC(=O)OCC
123-29-5
C11H22O2
186.295
MFCD00009570
BYEVBITUADOIGY-UHFFFAOYSA-N
ethyl pelargonate,nonanoic acid, ethyl ester,ethyl nonylate,wine ether,nonanoic acid ethyl ester,natural,ethyl n-nonanoate,unii-ksh683s98j,fema no. 2447,pelargonic acid ethyl ester
31251
CHEBI:87501
ethyl nonanoate
CCCCCCCCC(=O)OCC
Flash Point 94°C (201°F)
Melting Point -44°C
Odor Pleasant
Boiling Point 227°C to 229°C
Quantity 100mL
Linear Formula CH3(CH2)7CO2CH2CH3
Beilstein 1759169
Merck Index 14,3838
Formula Weight 186.3
Percent Purity 97%
Chemical Name or Material Ethyl nonanoate
Show More Show Less

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : RA6845000

TSCA : TSCA