Alfa Aesar™ Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate, 98+%

Manufacturer: Alfa Aesar™ H55470MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55470MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 38202-27-6
    Molecular Formula C13H19NO4S
    Formula Weight 285.36
    MDL Number MFCD00009244
    Beilstein 2144156
    Synonym O-(2-Mesitylenesulfonyl)acetohydroxamic acid ethyl ester
    Chemical Name or Material Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate

    Specifications

    Formula Weight 285.36
    Molecular Formula C13H19NO4S
    MDL Number MFCD00009244
    Beilstein 2144156
    Synonym O-(2-Mesitylenesulfonyl)acetohydroxamic acid ethyl ester
    Melting Point 54° to 56°C
    Quantity 250mg
    CAS 38202-27-6
    Recommended Storage 2° to 8°C
    Chemical Name or Material Ethyl O-(2-mesitylenesulfonyl)acetohydroxamate
    Assay ≥98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit