Alfa Aesar™ Ethyl o-tolylacetate, 98%

Manufacturer: Alfa Aesar™ A1195530

Quantity 250g

 View more versions of this product

Catalog No. AAA1195530

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 40291-39-2
    Molecular Formula C11H14O2
    Formula Weight 178.23
    MDL Number MFCD00009179
    Beilstein 879835
    Synonym o-Tolylacetic acid ethyl ester; Ethyl 2-methylphenylacetate
    Chemical Name or Material Ethyl o-tolylacetate

    Specifications

    Formula Weight 178.23
    Synonym o-Tolylacetic acid ethyl ester; Ethyl 2-methylphenylacetate
    Molecular Formula C11H14O2
    CAS 40291-39-2
    Quantity 250g
    MDL Number MFCD00009179
    Boiling Point 238° to 239°C
    Beilstein 879835
    Flash Point 106°C (222°F)
    Chemical Name or Material Ethyl o-tolylacetate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit