Alfa Aesar™ Ethyl oxanilate, 98%

Manufacturer: Alfa Aesar™ B2328506

Quantity 5g

 View more versions of this product

Catalog No. AAB2328506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1457-85-8
    Molecular Formula C10H11NO3
    Formula Weight 193.2
    MDL Number MFCD00009118
    Beilstein 2098703
    Synonym Oxanilic acid ethyl ester
    Chemical Name or Material Ethyl oxanilate

    Specifications

    Formula Weight 193.2
    Molecular Formula C10H11NO3
    MDL Number MFCD00009118
    CAS 1457-85-8
    Beilstein 2098703
    Synonym Oxanilic acid ethyl ester
    Melting Point 64° to 70°C
    Quantity 5g
    Chemical Name or Material Ethyl oxanilate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit