Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl p-Anisate 99.0+%, TCI America™

Click to view available options
Quantity:
25mL
100mL
500mL
94-30-4
C10H12O3
180.203
MFCD00015151
FHUODBDRWMIBQP-UHFFFAOYSA-N
ethyl p-anisate,ethyl anisate,ethyl p-methoxybenzoate,p-anisic acid, ethyl ester,benzoic acid, 4-methoxy-, ethyl ester,4-methoxybenzoic acid ethyl ester,benzoic acid, p-methoxy-, ethyl ester,ethyl p-anisoate,unii-kj95h2s7nm,p-anisic acid ethyl ester
60979
ethyl 4-methoxybenzoate
CCOC(=O)C1=CC=C(C=C1)OC
94-30-4
C10H12O3
180.203
MFCD00015151
FHUODBDRWMIBQP-UHFFFAOYSA-N
ethyl p-anisate,ethyl anisate,ethyl p-methoxybenzoate,p-anisic acid, ethyl ester,benzoic acid, 4-methoxy-, ethyl ester,4-methoxybenzoic acid ethyl ester,benzoic acid, p-methoxy-, ethyl ester,ethyl p-anisoate,unii-kj95h2s7nm,p-anisic acid ethyl ester
60979
ethyl 4-methoxybenzoate
CCOC(=O)C1=CC=C(C=C1)OC
Quantity 500mL
Color Colorless
Boiling Point 143°C
Formula Weight 180.20
Physical Form Clear Liquid at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material Ethyl p-Anisate

EINECSNumber : (3)-3054

RTECSNumber : BZ4697000

TSCA : Yes