Alfa Aesar™ Ethyl phenyl sulfone, 98+%

Manufacturer: Alfa Aesar™ A1071014

Quantity 25g

 View more versions of this product

Catalog No. AAA1071014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 599-70-2
    Molecular Formula C8H10O2S
    Formula Weight 170.23
    MDL Number MFCD00025081
    Beilstein 2045095
    Chemical Name or Material Ethyl phenyl sulfone

    Specifications

    Formula Weight 170.23
    MDL Number MFCD00025081
    Quantity 25g
    CAS 599-70-2
    Beilstein 2045095
    Molecular Formula C8H10O2S
    Melting Point 40° to 44°C
    Chemical Name or Material Ethyl phenyl sulfone
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit