Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenylcyanoacetate 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
4553-07-5
C11H11NO2
189.21
MFCD00001866
SXIRJEDGTAKGKU-UHFFFAOYNA-N
s-dibenzyl 2-aminopentanedioate hydrochloride,h-glu obzl-obzl.hcl,l-glutamic acid dibenzyl ester hydrochloride,l-glutamic acid, bis phenylmethyl ester, hydrochloride,s-dibenzyl 2-aminopentanedioate hcl,dibenzyl l-glutaminate hydrochloride,glutamic acid, dibenzyl ester, hydrochloride, l,1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride,h-glu obzl-obzl.hci,h-glu obzl-obzl hcl
199613
dibenzyl (2S)-2-aminopentanedioate;hydrochloride
CCOC(=O)C(C#N)C1=CC=CC=C1
4553-07-5
C11H11NO2
189.21
MFCD00001866
SXIRJEDGTAKGKU-UHFFFAOYNA-N
s-dibenzyl 2-aminopentanedioate hydrochloride,h-glu obzl-obzl.hcl,l-glutamic acid dibenzyl ester hydrochloride,l-glutamic acid, bis phenylmethyl ester, hydrochloride,s-dibenzyl 2-aminopentanedioate hcl,dibenzyl l-glutaminate hydrochloride,glutamic acid, dibenzyl ester, hydrochloride, l,1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride,h-glu obzl-obzl.hci,h-glu obzl-obzl hcl
199613
dibenzyl (2S)-2-aminopentanedioate;hydrochloride
CCOC(=O)C(C#N)C1=CC=CC=C1
Refractive Index 1.5033 to 1.5053
Boiling Point 275°C
Specific Gravity 1.09
Color Colorless to Yellow
Quantity 5g
Packaging Glass bottle
Density 1.0900g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C6H5CH(CN)CO2C2H5
Beilstein 09, 854
Formula Weight 189.21
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Ethyl phenylcyanoacetate, 97%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-921-4