Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Phenylpropiolate 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2216-94-6
C11H10O2
174.2
MFCD00009185
ACJOYTKWHPEIHW-UHFFFAOYSA-N
ethyl phenylpropiolate,ethylphenylpropiolate,ethyl 3-phenylpropiolate,ethyl phenylpropriolate,2-propynoic acid, 3-phenyl-, ethyl ester,ethyl 3-phenylpropynoate,ethyl phenylpropynoate,ethyl phenylacetylenecarboxylate,phenylpropiolic acid ethyl ester,ethyl 3-phenyl-2-propynoate
91516
ethyl 3-phenylprop-2-ynoate
CCOC(=O)C#CC1=CC=CC=C1
2216-94-6
C11H10O2
174.2
MFCD00009185
ACJOYTKWHPEIHW-UHFFFAOYSA-N
ethyl phenylpropiolate,ethylphenylpropiolate,ethyl 3-phenylpropiolate,ethyl phenylpropriolate,2-propynoic acid, 3-phenyl-, ethyl ester,ethyl 3-phenylpropynoate,ethyl phenylpropynoate,ethyl phenylacetylenecarboxylate,phenylpropiolic acid ethyl ester,ethyl 3-phenyl-2-propynoate
91516
ethyl 3-phenylprop-2-ynoate
CCOC(=O)C#CC1=CC=CC=C1
Density 1.0500g/mL
Refractive Index 1.5510 to 1.553
Boiling Point 260°C to 270°C
Flash Point >112°C
Specific Gravity 1.05
Quantity 25g
Color Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula C6H5C∽CCO2C2H5
Beilstein 09, 634
Formula Weight 174.2
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Ethyl phenylpropiolate, 98%
Show More Show Less

EINECSNumber : 218-703-8

RTECSNumber : UE0110000