Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl Phenylsulfonylacetate 98.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 44°C |
Color | White |
Quantity | 25 g |
Formula Weight | 228.26 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Ethyl Phenylsulfonylacetate |
Safety and Handling
TSCA : No