Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Picolinate 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
2524-52-9
C8H9NO2
151.16
MFCD00006292
FQYYIPZPELSLDK-UHFFFAOYSA-N
ethyl picolinate,ethyl 2-picolinate,ethyl 2-pyridinecarboxylate,picolinic acid, ethyl ester,2-picolinic acid ethyl ester,2-pyridinecarboxylic acid, ethyl ester,2-ethoxycarbonyl pyridine,picolinic acid ethyl ester,2-pyridinecarboxylic acid ethyl ester,pyridine-2-carboxylic acid ethyl ester
17307
ethyl pyridine-2-carboxylate
CCOC(=O)C1=CC=CC=N1
2524-52-9
C8H9NO2
151.16
MFCD00006292
FQYYIPZPELSLDK-UHFFFAOYSA-N
ethyl picolinate,ethyl 2-picolinate,ethyl 2-pyridinecarboxylate,picolinic acid, ethyl ester,2-picolinic acid ethyl ester,2-pyridinecarboxylic acid, ethyl ester,2-ethoxycarbonyl pyridine,picolinic acid ethyl ester,2-pyridinecarboxylic acid ethyl ester,pyridine-2-carboxylic acid ethyl ester
17307
ethyl pyridine-2-carboxylate
CCOC(=O)C1=CC=CC=N1
Melting Point 2°C
Boiling Point 240°C to 241°C
Flash Point 107°C
Specific Gravity 1.11
Refractive Index 1.5090 to 1.511
Infrared Spectrum Authentic
Quantity 25mL
Color Pink or Yellow
Packaging Glass bottle
Density 1.1100g/mL
Assay Percent Range 99%
Beilstein 22, 35
Solubility Information Solubility in water: miscible
Formula Weight 151.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Ethyl picolinate, 99%
Show More Show Less

EINECSNumber : 219-758-0

TSCA : TSCA