Learn More
Ethyl propionylacetate, 95%, Thermo Scientific Chemicals
Description
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl propionylacetate acts as a precursor used in the preparation of 1-ethyl- and 1-n-propyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid bearing one or two substituents on the benzene ring. Further, it is used to prepare 3-propionyl-2H-cyclohepta[b]furan-2-one.
Solubility
Miscible with glacial acetic acid.
Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
Specifications
Density | 1.012 |
Boiling Point | 190°C to 192°C |
Flash Point | 77°C (170°F) |
Refractive Index | 1.422 |
Quantity | 5 g |
Beilstein | 970270 |
Solubility Information | Miscible with glacial acetic acid. |
Formula Weight | 144.17 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl propionylacetate |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
EINECSNumber | 225-593-5 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.