Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-3-hydroxybutyrate, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
24915-95-5
C6H12O3
132.16
MFCD00075386
OMSUIQOIVADKIM-RXMQYKEDSA-N
ethyl r-3-hydroxybutyrate,ethyl 3r-3-hydroxybutanoate,ethyl r-3-hydroxybutanoate,ethyl r---3-hydroxybutyrate,r-ethyl 3-hydroxybutanoate,r---3-hydroxybutyric acid ethyl ester,r-3-hydroxybutyric acid ethyl ester,butanoic acid, 3-hydroxy-, ethyl ester, 3r,r---3-hydroxy-n-butyric acid ethyl ester
440030
CHEBI:28707
ethyl (3R)-3-hydroxybutanoate
CCOC(=O)CC(C)O
24915-95-5
C6H12O3
132.16
MFCD00075386
OMSUIQOIVADKIM-RXMQYKEDSA-N
ethyl r-3-hydroxybutyrate,ethyl 3r-3-hydroxybutanoate,ethyl r-3-hydroxybutanoate,ethyl r---3-hydroxybutyrate,r-ethyl 3-hydroxybutanoate,r---3-hydroxybutyric acid ethyl ester,r-3-hydroxybutyric acid ethyl ester,butanoic acid, 3-hydroxy-, ethyl ester, 3r,r---3-hydroxy-n-butyric acid ethyl ester
440030
CHEBI:28707
ethyl (3R)-3-hydroxybutanoate
CCOC(=O)CC(C)O
Boiling Point 75°C to 76°C (12.0mmHg)
Specific Gravity 1.017
Specific Rotation -45.5
Refractive Index 1.4195 to 1.4215
Density 1.0170g/mL
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Colorless
Specific Rotation Condition −45.5° (20°C 589nm,c=1,CHCl3)
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH(OH)CH2COOCH2CH3
Solubility Information Solubility in water: soluble in water
Formula Weight 132.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl (R)-3-hydroxybutyrate, 98%
Show More Show Less