Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
90866-33-4
C6H11ClO3
166.601
ZAJNMXDBJKCCAT-RXMQYKEDSA-N
ethyl r-+-4-chloro-3-hydroxybutyrate,r-ethyl 4-chloro-3-hydroxybutanoate,ethyl 3r-4-chloro-3-hydroxybutanoate,ethyl r-4-chloro-3-hydroxybutyrate,ethyl r-+-4-chloro-3-hydroxybutanoate,r-4-chloro-3-hydroxybutyric acid ethyl ester,s-4-chloro-3-hydroxy-butyric acid ethyl ester,r-4-chloro-3-hydroxy-n-butyric acid ethyl ester,ethyl r-4-chloro-3-hydroxybutanoate,butanoic acid, 4-chloro-3-hydroxy-, ethyl ester, 3r
2734445
ethyl (3R)-4-chloro-3-hydroxybutanoate
CCOC(=O)CC(CCl)O
90866-33-4
C6H11ClO3
166.601
ZAJNMXDBJKCCAT-RXMQYKEDSA-N
ethyl r-+-4-chloro-3-hydroxybutyrate,r-ethyl 4-chloro-3-hydroxybutanoate,ethyl 3r-4-chloro-3-hydroxybutanoate,ethyl r-4-chloro-3-hydroxybutyrate,ethyl r-+-4-chloro-3-hydroxybutanoate,r-4-chloro-3-hydroxybutyric acid ethyl ester,s-4-chloro-3-hydroxy-butyric acid ethyl ester,r-4-chloro-3-hydroxy-n-butyric acid ethyl ester,ethyl r-4-chloro-3-hydroxybutanoate,butanoic acid, 4-chloro-3-hydroxy-, ethyl ester, 3r
2734445
ethyl (3R)-4-chloro-3-hydroxybutanoate
CCOC(=O)CC(CCl)O
Packaging Glass bottle
Quantity 1g
Assay Percent Range 98%
Formula Weight 166.61
Chemical Name or Material Ethyl (R)-(+)-4-chloro-3-hydroxybutyrate