Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-4-cyano-3-hydroxybutanoate, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
141942-85-0
C7H11NO3
157.17
LOQFROBMBSKWQY-ZCFIWIBFSA-N
ethyl r---4-cyano-3-hydroxybutyate,ethyl r---4-cyano-3-hydroxybutyrate,ethyl 3r-4-cyano-3-hydroxybutanoate,r-4-cyano-3-hydroxybutyric acid ethyl ester,r-ethyl 4-cyano-3-hydroxybutyrate,ethyl-r-+-4-cyano-3-hydroxybutanoate,ethyl r-4-cyano-3-hydroxybutyrate,r-ethyl 4-cyano-3-hydroxybutanoate,r-4-cyano-3-hydroxy-butyric acid ethyl ester,r---4-cyano-3-hydroxybutyric acid ethyl ester
2733879
ethyl (3R)-4-cyano-3-hydroxybutanoate
CCOC(=O)CC(CC#N)O
141942-85-0
C7H11NO3
157.17
LOQFROBMBSKWQY-ZCFIWIBFSA-N
ethyl r---4-cyano-3-hydroxybutyate,ethyl r---4-cyano-3-hydroxybutyrate,ethyl 3r-4-cyano-3-hydroxybutanoate,r-4-cyano-3-hydroxybutyric acid ethyl ester,r-ethyl 4-cyano-3-hydroxybutyrate,ethyl-r-+-4-cyano-3-hydroxybutanoate,ethyl r-4-cyano-3-hydroxybutyrate,r-ethyl 4-cyano-3-hydroxybutanoate,r-4-cyano-3-hydroxy-butyric acid ethyl ester,r---4-cyano-3-hydroxybutyric acid ethyl ester
2733879
ethyl (3R)-4-cyano-3-hydroxybutanoate
CCOC(=O)CC(CC#N)O
Density 1.1140g/mL
Refractive Index 1.4460 to 1.4500
Infrared Spectrum Authentic
Specific Gravity 1.114
Quantity 1g
Boiling Point 270.0°C
Flash Point >190°C
Packaging Glass bottle
Specific Rotation -4.8° to -5.2° (20°C, 589nm) (neat)
Color Colorless to Yellow
Assay Percent Range 94% min. (GC)
Linear Formula NCCH2CH(OH)CH2COOCH2CH3
Solubility Information Solubility in water: soluble in water
Formula Weight 157.17
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Ethyl (R)-4-cyano-3-hydroxybutanoate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning