Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (R)-nipecotate L-tartarate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass Bottle
167392-57-6
C12H21NO8
307.299
MFCD00799538
HHPGQKZOPPDLNH-RATVZYDNSA-N
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate,3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate,3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
53249980
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
167392-57-6
C12H21NO8
307.299
MFCD00799538
HHPGQKZOPPDLNH-RATVZYDNSA-N
s,s-tartaric acid; ethyl 3r-piperidine-3-carboxylate,3s-2,3-bis oxidanyl butanedioic acid; ethyl 3r-piperidine-3-carboxylate,3s-2,3-dihydroxybutanedioic acid; 3r-3-piperidinecarboxylic acid ethyl ester
53249980
(3S)-2,3-dihydroxybutanedioic acid;ethyl (3R)-piperidine-3-carboxylate
CCOC(=O)C1CCCNC1.C(C(C(=O)O)O)(C(=O)O)O
Packaging Glass Bottle
Quantity 10g
Assay Percent Range 97%
Formula Weight 307.3
Chemical Name or Material Ethyl (R)-nipecotate L-tartarate, 97%

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.