Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl (S)-(-)-2-methoxypropionate, 98%, ACROS Organics™

Click to view available options
Quantity:
1mL
10mL
Packaging:
Glass bottle
41918-08-5
C6H12O3
132.16
MFCD01632590
WHRLOJCOIKOQGL-YFKPBYRVSA-N
ethyl 2s-2-methoxypropanoate,s-ethyl 2-methoxypropanoate,ethyl s-2-methoxypropanoate,ethyl s---2-methoxypropionate,2-methoxypropionic acid ethyl ester,s, ?-2-methoxypropionic acid ethyl ester,propanoic acid,2-methoxy-, ethyl ester, 2s
5702567
ethyl (2S)-2-methoxypropanoate
CCOC(=O)C(C)OC
41918-08-5
C6H12O3
132.16
MFCD01632590
WHRLOJCOIKOQGL-YFKPBYRVSA-N
ethyl 2s-2-methoxypropanoate,s-ethyl 2-methoxypropanoate,ethyl s-2-methoxypropanoate,ethyl s---2-methoxypropionate,2-methoxypropionic acid ethyl ester,s, ?-2-methoxypropionic acid ethyl ester,propanoic acid,2-methoxy-, ethyl ester, 2s
5702567
ethyl (2S)-2-methoxypropanoate
CCOC(=O)C(C)OC
Specific Rotation Condition − 89.00 (20.00°C c=neat)
Packaging Glass bottle
Color Colorless to Yellow
Quantity 1mL
Refractive Index 1.3970 to 1.3990
Infrared Spectrum Authentic
Flash Point >110°C
Specific Rotation − 89.00
Assay Percent Range 98%
Linear Formula CH3CH(OCH3)COOCH2CH3
Beilstein 03, III, 446
Formula Weight 132.16
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Ethyl (S)-(-)-2-methoxypropionate
Show More Show Less