Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Silicate, Condensed, Spectrum™

Click to view available options
Quantity:
3 kg
200 L
Packaging:
Amber Glass Bottle
Drum
78-10-4
C8H20O4Si
208.33
BOTDANWDWHJENH-UHFFFAOYSA-N
tetraethyl silicate
CCO[Si](OCC)(OCC)OCC
78-10-4
C8H20O4Si
208.33
BOTDANWDWHJENH-UHFFFAOYSA-N
tetraethyl silicate
CCO[Si](OCC)(OCC)OCC

E1105, 78-10-4

Spectrum™ Ethyl Silicate, Condensed is also known under the name Tetraethyl orthosilicate and is often abbreviated as TEOS. It is used as a crosslinking agent in silicone polymers and as a precursor to silicon dioxide in the semiconductor industry.
Packaging Amber Glass Bottle
Acidity 1e-005
Specific Gravity 0.930-0.940
Quantity 3 kg
CAS Max % 1
UN Number UN1292