Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl stearate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
111-61-5
C20H40O2
312.52
MFCD00009006
MVLVMROFTAUDAG-UHFFFAOYSA-N
ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate
8122
CHEBI:84936
ethyl octadecanoate
CCCCCCCCCCCCCCCCCC(=O)OCC
111-61-5
C20H40O2
312.52
MFCD00009006
MVLVMROFTAUDAG-UHFFFAOYSA-N
ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate
8122
CHEBI:84936
ethyl octadecanoate
CCCCCCCCCCCCCCCCCC(=O)OCC
Melting Point 30°C to 39°C
Packaging Glass bottle
Boiling Point 213°C to 215°C (15.0mmHg)
Flash Point >112°C
Color White
Quantity 5g
Infrared Spectrum Authentic
Appearance this product may appear as a very pale yellow liquid
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)16CO2C2H5
Beilstein 02, 379
Merck Index 15, 8930
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,acetone
Formula Weight 312.52
Physical Form Crystalline Mass
Percent Purity 97%
Chemical Name or Material Ethyl stearate, 97%
Show More Show Less

EINECSNumber : 203-887-4

RTECSNumber : WI3600000

TSCA : TSCA