Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl stearate, 97%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Specifications
Quantity | 25g |
Flash Point | >112°C |
Boiling Point | 213°C to 215°C (15.0mmHg) |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Melting Point | 30°C to 39°C |
Color | White |
Appearance | this product may appear as a very pale yellow liquid |
Assay Percent Range | 96% min. (GC) |
Linear Formula | CH3(CH2)16CO2C2H5 |
Show More |
Safety and Handling
EINECSNumber : 203-887-4
RTECSNumber : WI3600000
TSCA : TSCA