Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Sulfide 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
100mL
500mL
352-93-2
C4H10S
90.184
LJSQFQKUNVCTIA-UHFFFAOYSA-N
diethyl sulfide,ethyl sulfide,ethyl thioether,3-thiapentane,diethylthioether,thioethyl ether,ethylthioethane,1,1'-thiobisethane,diethyl sulphide,ethane, 1,1'-thiobis
9609
CHEBI:27710
ethylsulfanylethane
CCSCC
352-93-2
C4H10S
90.184
LJSQFQKUNVCTIA-UHFFFAOYSA-N
diethyl sulfide,ethyl sulfide,ethyl thioether,3-thiapentane,diethylthioether,thioethyl ether,ethylthioethane,1,1'-thiobisethane,diethyl sulphide,ethane, 1,1'-thiobis
9609
CHEBI:27710
ethylsulfanylethane
CCSCC
Quantity 100mL
Color Colorless
Infrared Spectrum Authentic
Density 0.83
Refractive Index 1.4410 to 1.4430 (20°C, 589nm)
Linear Formula (C2H5)2S
Beilstein 01, 344
Merck Index 15, 3905
Formula Weight 90.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Ethyl sulfide, 98%
Show More Show Less

STENCH

EINECSNumber : 206-526-9

RTECSNumber : LC7200000

TSCA : TSCA