Alfa Aesar™ Ethyl thiophene-2-carboxylate, 98+%

Manufacturer: Alfa Aesar™ A1230209

Quantity 10g

 View more versions of this product

Catalog No. AAA1230209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2810-04-0
    Molecular Formula C7H8O2S
    Formula Weight 156.2
    MDL Number MFCD00005436
    Beilstein 112678
    Synonym Thiophene-2-carboxylic acid ethyl ester
    Chemical Name or Material Ethyl thiophene-2-carboxylate

    Specifications

    Formula Weight 156.2
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Synonym Thiophene-2-carboxylic acid ethyl ester
    Health Hazard 1 Warning
    MDL Number MFCD00005436
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Quantity 10g
    CAS 2810-04-0
    Beilstein 112678
    Boiling Point 217° to 218°C
    Molecular Formula C7H8O2S
    Flash Point 88°C (190°F)
    Chemical Name or Material Ethyl thiophene-2-carboxylate
    Assay ≥98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit