Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl trans-2-amino-4-cyclohexene-1-carboxylate hydrochloride, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Specifications
Melting Point | 107°C to 109°C |
Color | Beige to White or Gray |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | C6H8(NH2)COOCH2CH3·HCl |
Quantity | 1 g |
Solubility Information | Solubility in water: .. Other solubilities: soluble in ethanol |
Formula Weight | 205.68 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction