Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl trans-3-(1-pyrrolidinyl)acrylate, 98+%, Thermo Scientific™

Click to view available options
Quantity:
1g
65651-80-1
C9H15NO2
169.22
MFCD01075735
VFFHLZJSZHWXAJ-UHFFFAOYSA-N
ethyl trans-3-1-pyrrolidinyl acrylate, e-ethyl 3-pyrrolidin-1-yl acrylate, ethyl 2e-3-pyrrolidin-1-yl prop-2-enoate, 3-pyrrolidin-1-yl-acrylic acid ethyl ester, hcnd@nadfu^u zfjjaap, 2-propenoic acid, 3-1-pyrrolidinyl-, ethyl ester, 2e, 3-pyrrolizinoacrylic acid ethyl ester, ethyl 2e-3-pyrrolidin-1-ylacrylate, ethyl 2e-3-pyrrolidinylprop-2-enoate, ethyl e-3-pyrrolidin-1-ylprop-2-enoate
639785
ethyl (E)-3-pyrrolidin-1-ylprop-2-enoate
CCOC(=O)C=CN1CCCC1
65651-80-1
C9H15NO2
169.22
MFCD01075735
VFFHLZJSZHWXAJ-UHFFFAOYSA-N
ethyl trans-3-1-pyrrolidinyl acrylate, e-ethyl 3-pyrrolidin-1-yl acrylate, ethyl 2e-3-pyrrolidin-1-yl prop-2-enoate, 3-pyrrolidin-1-yl-acrylic acid ethyl ester, hcnd@nadfu^u zfjjaap, 2-propenoic acid, 3-1-pyrrolidinyl-, ethyl ester, 2e, 3-pyrrolizinoacrylic acid ethyl ester, ethyl 2e-3-pyrrolidin-1-ylacrylate, ethyl 2e-3-pyrrolidinylprop-2-enoate, ethyl e-3-pyrrolidin-1-ylprop-2-enoate
639785
ethyl (E)-3-pyrrolidin-1-ylprop-2-enoate
CCOC(=O)C=CN1CCCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Melting Point 39°C to 43°C
Boiling Point 118°C (1mmHg)
Beilstein 4399757
Quantity 1g
Formula Weight 169.22
Percent Purity ≥98%
Chemical Name or Material Ethyl trans-3-(1-pyrrolidinyl)acrylate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Keep cold