Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl trans-4-bromocinnamate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
24393-53-1
C11H11BrO2
255.111
MFCD00012226
YOOKYIPLSLPRTC-VMPITWQZSA-N
ethyl trans-4-bromocinnamate,e-ethyl 3-4-bromophenyl acrylate,ethyl 3-4-bromophenyl acrylate,2-propenoic acid, 3-4-bromophenyl-, ethyl ester,ethyl 2e-3-4-bromophenyl prop-2-enoate,2-propenoic acid, 3-4-bromophenyl-, ethyl ester, 2e,ethyl p-bromocinnamate,ethyl e-4-bromocinnamate,ethyl-trans-4-bromocinnamate,4-bromocinnamic acid ethyl ester
688115
ethyl (E)-3-(4-bromophenyl)prop-2-enoate
CCOC(=O)C=CC1=CC=C(C=C1)Br
24393-53-1
C11H11BrO2
255.111
MFCD00012226
YOOKYIPLSLPRTC-VMPITWQZSA-N
ethyl trans-4-bromocinnamate,e-ethyl 3-4-bromophenyl acrylate,ethyl 3-4-bromophenyl acrylate,2-propenoic acid, 3-4-bromophenyl-, ethyl ester,ethyl 2e-3-4-bromophenyl prop-2-enoate,2-propenoic acid, 3-4-bromophenyl-, ethyl ester, 2e,ethyl p-bromocinnamate,ethyl e-4-bromocinnamate,ethyl-trans-4-bromocinnamate,4-bromocinnamic acid ethyl ester
688115
ethyl (E)-3-(4-bromophenyl)prop-2-enoate
CCOC(=O)C=CC1=CC=C(C=C1)Br
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Color Yellow
Refractive Index 1.5950 to 1.5970 (20°C, 589nm)
Density 1.3590g/mL
Assay Percent Range 98%
Formula Weight 255.11
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Ethyl trans-4-bromocinnamate
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.