Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyl tridecanoate, 97%

Click to view available options
Quantity:
5g
25g
100g
28267-29-0
C15H30O2
242.403
MFCD00008978
QJYYMNOTJXIOBP-UHFFFAOYSA-N
tridecanoic acid ethyl ester,tridecanoic acid, ethyl ester,ethyltridecanoate,ethyl=tridecanoate,tridecanoic acid, ethylester,acmc-1cr51,ethyl tridecanoate,n-tridecanoic acid ethyl ester
119908
ethyl tridecanoate
CCCCCCCCCCCCC(=O)OCC
28267-29-0
C15H30O2
242.403
MFCD00008978
QJYYMNOTJXIOBP-UHFFFAOYSA-N
tridecanoic acid ethyl ester,tridecanoic acid, ethyl ester,ethyltridecanoate,ethyl=tridecanoate,tridecanoic acid, ethylester,acmc-1cr51,ethyl tridecanoate,n-tridecanoic acid ethyl ester
119908
ethyl tridecanoate
CCCCCCCCCCCCC(=O)OCC
Flash Point >110°C (230°F)
Quantity 5g
Boiling Point 121°C to 123°C (2.2mmHg)
Linear Formula CH3(CH2)11CO2CH2CH3
Beilstein 1773737
Formula Weight 242.41
Percent Purity 97%
Chemical Name or Material Ethyl tridecanoate