Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Ethyl tridecanoate, 97%
Click to view available options
Quantity:
5g
25g
100g
Specifications
Flash Point | >110°C (230°F) |
Density | 0.86 |
Quantity | 100g |
Refractive Index | 1.434 |
Boiling Point | 121°C to 123°C (2.2mmHg) |
Linear Formula | CH3(CH2)11CO2CH2CH3 |
Beilstein | 1773737 |
Formula Weight | 242.41 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl tridecanoate |
Safety and Handling
EINECSNumber : 248-929-2
TSCA : Yes
Recommended Storage : Ambient temperatures